Methyl orange 0,05% aqueous solution Indicator_pH

Supplier: Hach

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium salt, Methyl orange sodium salt, Helianthine

14832
HACH14832EA 21.1 GBP
HACH14832
Methyl orange 0,05% aqueous solution Indicator_pH
Methyl orange

pH Indicator for acidity titrations


0.50 g/L, APHA formulation. Color change pink (pH 3.2) to yellow (pH 4.4).

Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
Boiling Pt: 90
Flash Pt: 38
Storage Temperature: Ambient
MDL Number: MFCD00007502
CAS Number: 547-58-0
EINECS: 208-925-3
Merck Index: 13,06127

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR