(S)-(-)-β-Citronellol 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (S)-(-)-3,7-Dimethyl-6-octen-1-ol, (S)-(-)-ß-Citronellol, (S)-(-)-Citronellol, (-)-beta-Citronellol, (-)-β-Citronellol, (S)-3,7-Dimethyloct-6-en-1-ol, beta-Rhodinol

H60174.03 H60174.06
H60174.03EA 39.4 GBP
H60174.03 H60174.06
(S)-(-)-β-Citronellol 97
(S)-(-)-β-Citronellol
Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 225…226 °C
Density: 0.856
Flash Pt: 99 °C (210 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063214
CAS Number: 7540-51-4
EINECS: 231-415-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR