1-Benzylpyrrole 96%

Supplier: Thermo Fisher Scientific
Warning

H55936.03 H55936.06 H55936.14
H55936.03EA 28.5 GBP
H55936.03 H55936.06 H55936.14
1-Benzylpyrrole 96%
1-Benzylpyrrole
Formula: C₁₁H₁₁N
MW: 157.22 g/mol
Boiling Pt: 90 °C (0.9 mmHg)
Density: 1.020
Storage Temperature: Ambient
MDL Number: MFCD00963301
CAS Number: 2051-97-0
EINECS: 218-137-1
UN: 1993
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR