tert-Butyl glycolate 94%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: tert-Butyl 2-hydroxyacetate

H35711.03
H35711.03EA 475 GBP
H35711.03
tert-Butyl glycolate 94%
tert-Butyl glycolate

Purity: 94% CAS No.: 50595-15-8 Molecular Formula: C6H12O3 Formula Weight: 132.16 Storage Temperature: 2°C to 8°C Physical Form: Low melting solid Appearance: Colourless to yellow MDL No.: MFCD03452739

Formula: C₆H₁₂O₃
MW: 132.159 g/mol
MDL Number: MFCD03452739
CAS Number: 50595-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR