Methyl-6-amino-5-bromonicotinate 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Methyl 6-amino-5-bromonicotinate

H33991.06
H33991.06EA 0 GBP
H33991.06
Methyl-6-amino-5-bromonicotinate 98%
Methyl 6-amino-5-bromonicotinate

Purity: 98% CAS No.: 180340-70-9 Molecular Formula: C7H7BrN2O2 Formula Weight: 231.05 Melting Point: 166-169° MDL No.: MFCD06659561

Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD06659561
CAS Number: 180340-70-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR