Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Supplier: Avantor Performance Materials
H296-03EA 434 GBP
H296-03
Macron Fine Chemicals™ StandARd®, Platinum Cobalt Colour Standard (APHA 500)
Standards Colour Reference and Spectroscopy Standards
Corrosive.
APHA 500 contains 0,13% chloroplatinic acid, 0,11% cobalt (II) chloride, and 4,6% hydrogen chloride in water.