Ethyl cellulose

Supplier: Thermo Fisher Scientific

Synonyms: Ethylcellulose

H27621.MDEA 87.2 GBP
H27621.MD
Ethyl cellulose
Ethyl cellulose

Ethyl cellulose is used as a food additive and as an emulsifier. It is also used as a film-former in coatings, hot-melt adhesives and transfer inks. Further, it is employed as a binder, filler, plastic coating and diluents of color. In addition to this, it is used in lacquers and plastic toughing.


CAS No.: 9004-57-3 Refractive Index: 1.47 MDL No.: MFCD00131037

Density: 1.14
Storage Temperature: Ambient
MDL Number: MFCD00131037
CAS Number: 9004-57-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR