N-Methylallylamine 96%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N-Allylmethylamine

H27551.06 H27551.03
H27551.06EA 107 GBP
H27551.06 H27551.03
N-Methylallylamine 96%
N-Methylallylamine

Purity: 96% CAS No.: 627-37-2 Molecular Formula: CH3NHCH2CH=CH2 Formula Weight: 71.12 Storage Temperature: 2°C to 8°C Flash Point: -8°(-18°F) Refractive Index: 1.412 Boiling Point: 61-62° UN No.: 2733 MDL No.: MFCD00008641

Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 61…62 °C
Density: 0.748
Flash Pt: –8 °C (–18 °F)
MDL Number: MFCD00008641
CAS Number: 627-37-2
EINECS: 210-996-0
UN: 3286
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR