N-Acetyl-DL-leucine ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: N-Acetyl-DL-leucine, Ac-DL-Leu-OH, 2-Acetamido-4-methylpentanoic acid

H27441.22 H27441.14 H27441.06
H27441.22EA 342 GBP
H27441.22 H27441.14 H27441.06
N-Acetyl-DL-leucine ≥99%
N-Acetyl-DL-leucine

Purity: 99% CAS No.: 99-15-0 Molecular Formula: C8H15NO3 Formula Weight: 173.21 Storage Temperature: 2°C to 8°C Melting Point: 159-161° MDL No.: MFCD00026498

Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
Melting Pt: 159…161 °C
MDL Number: MFCD00026498
CAS Number: 99-15-0
EINECS: 202-734-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR