Sapphire Pipette Tips and Filter Tips
Supplier: Greiner Bio-One
778353_3840 772353_5760 774353_5760 776354_4800 776353_5760 778350_5000 778364_4800 773353_5760 773363_5760 771353_5760 775363_5760 775353_960 775350_15000 771350_10000 772350_5000 775364_4800 776350_10000 777350_5000 772354_4800 775354_4800 778354_4800 771354_4800 771361_5760 771363_5760 771364_4800 772361_5760 772364_4800 775361_5760 776361_5760 776364_4800 777354_4800 777361_3840 777364_4800 778361_3840 772351_5760 775351_5760 775355_4800 778351_3840 778355_4800 775362_5760 771355_4800 771365_4800 772355_4800 772365_4800 775365_4800 777355_4800 777365_4800 778365_4800 771351_5760 771352_5760 771362_5760 772352_5760 772362_5760 775352_5760 776351_5760 776352_5760 776355_4800 776362_5760 776365_4800 777351_3840 777352_3840 777362_3840 778352_3840 778362_3840
GREI778353_3840EA 764 GBP
GREI778353_3840 GREI772353_5760 GREI774353_5760 613-7369 GREI776353_5760 GREI778350_5000 GREI778364_4800 GREI773353_5760 GREI773363_5760 GREI771353_5760 GREI775363_5760 GREI775353_960 GREI775350_15000 GREI771350_10000 GREI772350_5000 GREI775364_4800 GREI776350_10000 GREI777350_5000 613-7363 613-7366 613-7375 613-7360 GREI771361_5760 GREI771363_5760 GREI771364_4800 GREI772361_5760 GREI772364_4800 GREI775361_5760 GREI776361_5760 GREI776364_4800 613-7372 GREI777361_3840 GREI777364_4800 GREI778361_3840 613-7361 613-7364 GREI775355_4800 613-7373 GREI778355_4800 GREI775362_5760 GREI771355_4800 GREI771365_4800 GREI772355_4800 GREI772365_4800 GREI775365_4800 GREI777355_4800 GREI777365_4800 GREI778365_4800 613-7358 613-7359 GREI771362_5760 613-7362 GREI772362_5760 613-7365 613-7367 613-7368 GREI776355_4800 GREI776362_5760 GREI776365_4800 613-7370 613-7371 GREI777362_3840 613-7374 GREI778362_3840
Sapphire Pipette Tips and Filter Tips
Pipette Tips
Flexibility is critical in terms of modern liquid handling solutions. This is why the Sapphire tips are manufactured in eight different sizes in a volume range from 10 to 1250 µl, including an extended length 10 µl tip for the recovery of small sample volumes. All our tips are made of medical grade polypropylene and are available as standard, low-retention, filter, or low-retention filter tips.
  • Fully automated production and packaging
  • Universal fit for all common pipettes
  • Tips (without filter) and racks are autoclavable
  • Filters manufactured without
More Product Information