Plasma collection tubes, with heparin
Supplier: Greiner Bio-One
454008_1200 454029_1200 454083_1200 454084_1200 454089_1200 455051_1200 455083_1200 455084_1200 456083_1200 456084_1200 454051_1200 454237_1200
GREI454008_1200EA 249 GBP
GREI454008_1200 GREI454029_1200 GREI454083_1200 GREI454084_1200 GREI454089_1200 720-2786 GREI455083_1200 720-2787 GREI456083_1200 GREI456084_1200 GREI454051_1200 GREI454237_1200
Plasma collection tubes, with heparin
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, clear.
Determinations in heparinised plasma for clinical chemistry. The use of a vacuum system eliminates the possibility of back flow occuring during blood collection.