Tubing, silicone, Sani-Tech®, ULTRA-C Series
Supplier: Saint Gobain Life Sciences
ULTRA-C-012-0 ULTRA-C-020-0 ULTRA-C-025-0 ULTRA-C-030-0 ULTRA-C-030-2 ULTRA-C-040-0 ULTRA-C-058-0 ULTRA-C-062-1 ULTRA-C-062-2 ULTRA-C-062-3 ULTRA-C-062-4 ULTRA-C-062-5 ULTRA-C-078-1 ULTRA-C-078-2 ULTRA-C-078-3 ULTRA-C-078-5 ULTRA-C-093-1 ULTRA-C-093-2 ULTRA-C-093-3 ULTRA-C-093-4 ULTRA-C-1000-4 ULTRA-C-125-1 ULTRA-C-125-2 ULTRA-C-125-3 ULTRA-C-125-4 ULTRA-C-125-5 ULTRA-C-156-4 ULTRA-C-187-1 ULTRA-C-187-2 ULTRA-C-187-3 ULTRA-C-187-4 ULTRA-C-187-5 ULTRA-C-250-1 ULTRA-C-250-2 ULTRA-C-250-3 ULTRA-C-250-4 ULTRA-C-312-1 ULTRA-C-312-2 ULTRA-C-312-3 ULTRA-C-312-4 ULTRA-C-312-5 ULTRA-C-375-2 ULTRA-C-375-3 ULTRA-C-375-4 ULTRA-C-375-5 ULTRA-C-500-2 ULTRA-C-500-3 ULTRA-C-500-4 ULTRA-C-500-5 ULTRA-C-625-2 ULTRA-C-625-4 ULTRA-C-625-5 ULTRA-C-750-4 ULTRA-C-750-5 ULTRA-C-750-6 ULTRA-C-875-2 ULTRA-C-875-4 ULTRA-C-875-5 ULTRA-C-875-6
GESSULTRA-C-012-0EA 44.8 GBP
GESSULTRA-C-012-0 GESSULTRA-C-020-0 GESSULTRA-C-025-0 GESSULTRA-C-030-0 GESSULTRA-C-030-2 GESSULTRA-C-040-0 GESSULTRA-C-058-0 GESSULTRA-C-062-1 GESSULTRA-C-062-2 GESSULTRA-C-062-3 GESSULTRA-C-062-4 GESSULTRA-C-062-5 GESSULTRA-C-078-1 GESSULTRA-C-078-2 GESSULTRA-C-078-3 GESSULTRA-C-078-5 GESSULTRA-C-093-1 GESSULTRA-C-093-2 GESSULTRA-C-093-3 GESSULTRA-C-093-4 GESSULTRA-C-1000-4 GESSULTRA-C-125-1 GESSULTRA-C-125-2 GESSULTRA-C-125-3 GESSULTRA-C-125-4 GESSULTRA-C-125-5 GESSULTRA-C-156-4 GESSULTRA-C-187-1 GESSULTRA-C-187-2 GESSULTRA-C-187-3 GESSULTRA-C-187-4 GESSULTRA-C-187-5 GESSULTRA-C-250-1 GESSULTRA-C-250-2 GESSULTRA-C-250-3 GESSULTRA-C-250-4 GESSULTRA-C-312-1 GESSULTRA-C-312-2 GESSULTRA-C-312-3 GESSULTRA-C-312-4 GESSULTRA-C-312-5 GESSULTRA-C-375-2 GESSULTRA-C-375-3 GESSULTRA-C-375-4 GESSULTRA-C-375-5 GESSULTRA-C-500-2 GESSULTRA-C-500-3 GESSULTRA-C-500-4 GESSULTRA-C-500-5 GESSULTRA-C-625-2 GESSULTRA-C-625-4 GESSULTRA-C-625-5 GESSULTRA-C-750-4 GESSULTRA-C-750-5 GESSULTRA-C-750-6 GESSULTRA-C-875-2 GESSULTRA-C-875-4 GESSULTRA-C-875-5 GESSULTRA-C-875-6
Tubing, silicone, Sani-Tech®, ULTRA-C Series
Tubing Transfer Tubing
Sani-Tech® ultra-pure platinum-cured silicone tubing meets the stringent requirements of the biopharmaceutical market.
  • Maximum temperature range: –62 to 260 °C (–80 to +500 °F)
  • Imparts no taste or odour to critical stream
  • Available in 50 durometer
  • Laser etched for lot traceability
It features ultra low extractables, excellent flexibility, longer pump life and non-yellowing formulation.
Applications include sterile fill and transfers, drug delivery, cell harvest, media processing, sanitary peristaltic pump tubing, production filtration andMore Product Information