Restriction enzymes, PstI, FastDigest™, Fermentas

Supplier: Thermo Fisher Scientific

FD0614 FD0615
FD0614EA 74.7 GBP
FD0614 FD0615
Restriction enzymes, PstI, FastDigest™, Fermentas
Nucleic Acid Reagents Restriction Enzymes

Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.


  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications


Designed to complete digestion in 5 to 15 minutes.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name PstI
Source
Overhang type 3' overhang
Compatible buffer FastDigest Green Buffer or FastDigest Buffer (colorless)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR