Actero™ Elite enrichment media
Supplier: FOODCHEK SYSTEMS
FCM-011 FCM-022 FCM-023 FCM-203 FCM-204
710-1615EA 265 GBP
710-1615 710-1617 FCHKFCM-023 FCHKFCM-203 FCHKFCM-204
Actero™ Elite enrichment media
Media for Microbiology Dehydrated Media for Microbiology
Food and environmental samples require enrichment before pathogen testing to amplify or grow any existing bacterial pathogens. Actero™ ELITE Enrichment Media works with a number of 'food pathogen testing systems' including lateral flow, ELISA and PCR platforms. For a custom solution, the Actero™ MediaPouch is for laboratories that require portion-packed media to use in automated media preparation instruments. It is available in pouches and can be custom packaged to meet your specific needs. Patent Pending Actero™ ELITE Enrichment Media represent enhanced formulations that further reduce the required enrichment time to an industry-leading low.
  • Different packages available
  • Robust and versatile