Coomassie® brilliant blue R-250, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor

Synonyms: Serva Blue R, Brillant blue R, N-[4-[[4-[(4-Ethoxyphenyl)amino]phenyl][4-[ethyl[(3-sulphophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulpho-benzenemethanaminium inner salt, sodium salt (1:1), Coomassie Blue R 250, Brilliant Cyanine 6B, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]benzhydrylene]cyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, Acid Blue 83, Brilliant Blue R250, Brilliant Blue R, Brilliant indocyanine 6B

F792-01EA 0 GBP
F792-01
Coomassie® brilliant blue R-250, BAKER, Laboratory Reagent, J.T.Baker®
Coomassie® Brilliant Blue R-250
Formula: C₄₅H₄₄N₃NaO₇S₂
MW: 825.98 g/mol
Melting Pt: 174…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00041762
CAS Number: 6104-59-2
EINECS: 228-060-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR