Supelco® Organic Reference Standard, D-(−)-Fructose

Supplier: Merck

F2793-500MG
F2793-500MGEA 18.8 GBP
F2793-500MG
Supelco® Organic Reference Standard, D-(−)-Fructose
Standards Organic Standards
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c22ea2691d99166878c994dbbca9338171d4fdb&vid=7e472086372b9cde6a816feddd4e40250ad4aacc&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) ᴅ-Levulose; Fruit sugar
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 57-48-7
MDL No. MFCD00148910
Molecular weight 180,16
Melting point 119 - 122 °C (dec.) (lit.)
Shipping conditions STD
Storage conditions RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR