Formaldehyde 37% w/w water Stabilised

Supplier: Honeywell Chemicals
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilisation: 10-15% MeOH

F1635-25ML F1635-500ML F1635-4L
F1635-25MLEA 38.2 GBP
F1635-25ML F1635-500ML F1635-4L
Formaldehyde 37% w/w water Stabilised
Formaldehyde
Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 93…96 °C (1.013 hPa)
Melting Pt: <–15 °C
Density: 1.080…1.090 g/cm³ (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8 III
Merck Index: 13,04259

Order Now

Specification Test Results

Vapor Density 1.03 (vs air)
Assay 37 - 38 %
Free acid (as HCOOH) Max. 0.03 %
Methanol (GC) 10 - 15 Vol.%
APHA Max. 10
Autoignition Temperature 572 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR