Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm

Supplier: Avantor
EXL-1211-1002UEA 0 GBP
EXL-1211-1002U EXL-1211-1003U EXL-1211-1046U EXL-1211-1246U EXL-1211-1502U EXL-1211-1503U EXL-1211-1546U EXL-1211-2046U EXL-1211-0202U EXL-1211-2502U EXL-1211-2503U EXL-1211-2546U EXL-1211-0302U EXL-1211-0346U EXL-1211-3546U EXL-1211-0503U EXL-1211-0546U EXL-1211-7503U EXL-1211-7546U EXL-1211-0502U
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
Avantor® ACE® Excel® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.

  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1,7 to 10 µm) and complementary phases
  • Method validation kit available containing 3 columns from different batches

Maximum recommended pH: 1,5 to 11,5

Max. pressure: 1000 bar
Order Now

SPECIFICATIONS

Environmentally Preferable
Carbon load 14,8%
Endcapped Yes
Max. pressure 1000 bar
pH range 1,5 - 11,5
Surface area 400 m²/g
USP code L1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR