pH/mV/°C meter, bench top, Eutech pH 2700
Supplier: EUTECH
ECPH270042GS
EUTEECPH270042GSEA 1180 GBP
EUTEECPH270042GS
pH/mV/°C meter, bench top, Eutech pH 2700
Electrochemistry Meters pH and Multi-Parameter Meters
This bench meter comes with a large, one-glance-sees-all screen. Users can view pH or ORP readings, with temperature, electrode status, calibration points, date and time all at once.
  • Up to 6-point push button calibration with auto-buffer recognition
  • Quick, easy electrode diagnosis with pH slope and offset display
  • Non volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Cal-due alarm – no more out-dated calibrations
  • Auto-logging function for convenient continuous monitoring
  • Splashproof housing and keypad protects against
More Product Information