Restriction enzymes, NotI, 10 U/µl, Fermentas

Supplier: Thermo Fisher Scientific

ER0591 ER0592 ER0595
ER0591EA 76 GBP
ER0591 ER0592 ER0595
Restriction enzymes, NotI, 10 U/µl, Fermentas
Nucleic Acid Reagents Restriction Enzymes

NotI restriction enzyme recognises GC^GGCCGC sites and cuts best at 37 °C in O buffer.


  • Superior quality, stringent quality control and industry leading manufacturing process
  • Convenient colour-coded five buffer system
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities


Applications: Molecular cloning, restriction site mapping, genotyping, southern blotting, restriction fragment length polymorphism (RFLP), SNP.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name NotI
Source
Overhang type 5' overhang
Compatible buffer FastDigest Green Buffer or FastDigest Buffer (colorless)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR