Restriction enzymes, HindIII, 10 U/μl, Fermentas

Supplier: Thermo Fisher Scientific

ER0501 ER0502 ER0505
ER0501EA 41.3 GBP
ER0501 ER0502 ER0505
Restriction enzymes, HindIII, 10 U/μl, Fermentas
Nucleic Acid Reagents Restriction Enzymes

HindIII restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions.


  • Convenient color-coded Five Buffer System
  • It includes universal Tango buffer for double-digestions
  • Reaction buffers contain premixed BSA
  • Wide selection of restriction endonuclease specificities


Recognises A^AGCTT sites and cuts best at 37 °C.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Enzyme name Hind III
Source
Overhang type 5' overhang
Compatible buffer FastDigest Green Buffer or FastDigest Buffer (colorless)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR