Jars, wide-mouth, with open top cap
Supplier: EP SCIENTIFIC
S320-0060 S320-0125 S321-0250 S340-0120 S120-0060 S120-0125 S121-0250
EPSCS320-0060EA 275 GBP
EPSCS320-0060 EPSCS320-0125 EPSCS321-0250 EPSCS340-0120 EPSCS120-0060 EPSCS120-0125 EPSCS121-0250
Jars, wide-mouth, with open top cap
Jars General Use Jars
Type III soda-lime glass
  • Designed for water sample collection for volatile organic analysis (VOA) and soil samples
  • Caps assembled along with bottles
  • Amber jars are recommended for light-sensitive analytes
  • With open-top white PP cap bonded with PTFE-lined silicone septum
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified jars and come with all the features of certified jars, but jars are not barcoded and CofA is not included.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement