Electroporators, Multiporator®
Supplier: EPPENDORF
4308031.000
EPPE4308031.000EA 0 GBP
EPPE4308031.000
Electroporators, Multiporator®
Electroporation Systems Electroporators
The Multiporator® is designed for the electroporation of eukaryotic cells. Applications include the transfection of animal and human cell lines, embryonic stem cells, primary cells, oocytes and plant cells.
  • Compact and easily portable
  • Easy to operate with clear, user-friendly display
  • Directly adjustable voltage and time constants
  • Built-in cuvette chamber for greater safety
  • Data documentation via printer or PC
  • Exponentially diminishing, electronically controlled pulse form
Specially formulated electroporation buffers are mycoplasma-tested and sterile, pyrogen- and endotoxin-free.
Order Now
SPECIFICATIONS
Module
Environmentally Preferable
Time constant
Voltage range
Multiple pulsing