Robust Temperature Data Logger, EBI 12-T237
Supplier: EBRO
1340-6658 1340-6637 1340-6601-0100
EBRO1340-6658EA 2530 GBP
EBRO1340-6658 EBRO1340-6637 EBRO1340-6601-0100
Robust Temperature Data Logger, EBI 12-T237
Data Loggers
The EBI 12-T237 temperature data logger is ideal for versatile routine checks in the food and pharmaceutical industries. With an extended measuring range, high-quality PT1000 sensor, IP 68 protection class and the option of use in autoclaves, it ensures reliable pasteurisation control and food safety.
  • Wide measuring range of –40 to +140 °C
  • High-quality PT1000 sensor (class A)
  • Rigid, external temperature sensor, Ø 3 mm, 100 mm long
  • Robust IP 68 protection class: waterproof and vapour-tight
This data logger was developed for routine checks in the foodMore Product Information