Safety underbench cabinets, type 90, UTS ergo line XL, 600 mm depth
Supplier: DUPERTHAL
29-061469-505 29-061469-005 29-061469-405 29-061467-005 29-061467-405 29-061467-505
DUEH29-061469-505EA 3310 GBP
DUEH29-061469-505 DUEH29-061469-005 DUEH29-061469-405 DUEH29-061467-005 135-1906 DUEH29-061467-505
Safety underbench cabinets, type 90, UTS ergo line XL, 600 mm depth
Furniture Storage Cabinets
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position andMore Product Information