Safety storage cabinets for toxic substances
Supplier: DUPERTHAL
20-201045-011
DUEH20-201045-011EA 1390 GBP
DUEH20-201045-011
Safety storage cabinets for toxic substances
Furniture Storage Cabinets
Safety storage cabinet ideal for storage of non flammable substances.
  • Torsion resistant wing doors with box gusset
  • Sheet steel cabinet with light grey colour RAL 7035
  • High quality safety lock for increased protection against break-ins
  • Four adjustable feet to compensate for uneven ground
  • Height adjustable perforated shelves for optimal ventilation
Cabinet features include wing door classical technology, integrated bracket for poison book with depth of 15 mm, bottom tray galvanised with material thickness 3 mm and ventilation via connection socket (NW 75More Product Information