Digital storage oscilloscopes
Supplier: DIDALAB
PMD100010_UK PMD100020_UK PMD100030_UK PMD100040_UK
DIDAPMD100010_UKEA 0 GBP
DIDAPMD100010_UK DIDAPMD100020_UK DIDAPMD100030_UK DIDAPMD100040_UK
Digital storage oscilloscopes
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
A range of digital storage oscilloscopes with bandwidth from 150 to 500 MHz.
  • Record length: 25000 points per channel
  • Display technology is VPO (visual persistence oscilloscope)
  • Split screen option for displaying each channel independently
  • Time base range: 1 nanosecond to 100 seconds/division
  • Offset position range: 2 mV/div ~ 100 mV/div : ±0,5 V; 200 mV/div ~ 5 V/div : ±25 V
Order Now
SPECIFICATIONS
Environmentally Preferable