Microtube racks, incubation
Supplier: DELTALAB
M-022
DELTM-022EA 205 GBP
DELTM-022
Microtube racks, incubation
Racks Tube Racks
This floating rack, made of PP, can incubate up to 24 microtubes in a water bath. Tubes are held snugly and then released when the rack is pressed
down on a benchtop.
  • Alphanumerically labelled for better sample identification
  • Autoclavable
  • Excellent chemical resistance