Sample container, with screw cap
Supplier: DELTALAB
409703
DELT409703EA 193 GBP
DELT409703
Sample container, with screw cap
Sample Containers General Purpose Containers
Ultra clear PP container with PE leak proof screw cap.
  • Moulded graduation in 20 ml increments up to 100 ml
  • Frosted panel on side for easy writing
  • Good chemical resistance
  • Good temperature resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable