DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) 99 HPCE_ASSAY_METHOD, Solid

Supplier: SIGMA ALDRICH MICROSCOPY

D0815-100MG
D0815-100MGEA 104 GBP
D0815-100MG
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) 99 HPCE_ASSAY_METHOD, Solid
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide)

DASPEI has been used for the labelling of hair cells in larval zebrafish. It accumulates in the mitochondria and has a large fluorescence stokes shift. It is also used for the relative measurements of mitochondrial membrane potential. Suitable as a mitochondrial stain in live cells. Storage temperature is 2 to 8 °C.

Formula: C₁₇H₂₁IN₂
MW: 380.273 g/mol
MDL Number: MFCD00011994
CAS Number: 3785-01-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd3dcc336d3543f8cace1c91c471f9ad7aefe164&vid=3c8de1b35f7053f58552ed9641c06f653f5423ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Red
Appearance (Form) Powder
Proton NMR spectrum Conforms to Structure
Solubility (Colour) Orange
Solubility (Turbidity)
0.5 mg/ml, DMSO
Clear
EmM
in MeOH
36.0 - 41.0
Wavelength 457 - 463 nm
Purity (HPLC) ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR