Supelco® TraceCERT® Organic Reference Standard, Disulfoton-sulfone

Supplier: Merck

CRM69056-50MG
CRM69056-50MGEA 134 GBP
CRM69056-50MG
Supelco® TraceCERT® Organic Reference Standard, Disulfoton-sulfone
Standards Organic Standards

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.


Caution: P264 - P270 - P301 + P310 - P405 - P501

Order Now

SPECIFICATIONS

Environmentally Preferable
M-Clarity quality level MQ300
CAS No. 2497-06-5
MDL No. MFCD00155401
Molecular weight 306,40
Hazard class description H300
Shipping conditions WICE
Storage conditions 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR