Freezing Box
Supplier: E3 Cortex
CARTISO15L
CORTCARTISO15LEA 205 GBP
CORTCARTISO15L
Freezing Box
Boxes Cooling/Freezing Boxes and Bags
Isothermal box made of rigid polyurethane (PU) foam is allowing the shipment in low temperature (refrigeration or freezing) of products sensitive to thermal variations.
  • Requires the supply of a suitable source of cold, in sufficient quantity
It is particularly suitable for the transfer of products to maintain positive low temperature between 1 to 8 °C or negative less than -20 °C between 48 to 120 h.
Cooling source (solid carbon dioxide or bag or plate of eutectic gel) available in variable quantity according to the constraints relating to theMore Product Information