Transport boxes, Biotainer
Supplier: E3 Cortex
3BIO000924
CORT3BIO000924EA 479 GBP
CORT3BIO000924
Transport boxes, Biotainer
Boxes Transport Boxes
These triple packaging designed for transporting all types of category A hazardous materials (UN 2814 / UN 2900 – class 6.2) at room temperature.
Certifications: Complies with regulation in force (IATA, IMDG, RID, ADR).