Bottle top dispensers, Corning®
Supplier: Corning
6840 6841 6842 6843 6844 6845
CORN6840EA 377 GBP
CORN6840 CORN6841 CORN6842 CORN6843 CORN6844 CORN6845
Bottle top dispensers, Corning®
Bottle Top Dispensers
Corning® bottle top dispensers are volumetric instruments designed for convenient and safe dispensing of liquids with no loss of reagent. Depending on the model, they can accommodate volumes from 0,25 to 100 ml.
  • Accurate and reproducible volume setting
  • Fully autoclavable for protection against contamination
  • Rotating valve block for convenient dispensing
  • Convenient 3-position discharge tube for dispensing, sample recovery, and emptying
  • Unique drip-free system for user safety at work
  • Easy in-lab calibration with built-in recalibration mechanism