Cleanroom bond papers, Munising LP®
Supplier: CONNECTICUT CLEANROOM
PB2S001BC PB2S002 PB3S001 PB3S002
CONNPB2S001BCEA 454 GBP
CONNPB2S001BC CONNPB2S002 CONNPB3S001 CONNPB3S002
Cleanroom bond papers, Munising LP®
Paper
Munising LP® bond papers are saturated with synthetic polymers and are free of inorganic fillers which prevents the doping of products and accepts standard printing inks without skipping, smudging or smearing.
  • Natural latex free
  • Heat resistant for use in laser printers and photocopy machines
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable