Conductivity standards, Supelco®
Supplier: Merck
COND147-1L COND50-500ML COND1412-1L COND1412-500ML COND147-500ML COND25-500ML COND75-500ML COND2060-500ML
COND147-1LEA 117 GBP
COND147-1L COND50-500ML COND1412-1L COND1412-500ML COND147-500ML COND25-500ML COND75-500ML COND2060-500ML
Conductivity standards, Supelco®
Standards Conductivity Standards
100 µmhos/cm = 100 µS/cm
Certifications: These certified reference materials (CRMs) ares produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.