Needle-Free Fluid Transfer System, CODAN Spikes / MicroSpikes
Supplier: CODAN
165260 16.5292 16.5264 165274 79.3800 79.3801
CODA165260EA 4950 GBP
CODA165260 CODA16.5292 CODA16.5264 CODA165274 CODA79.3800 CODA79.3801
Needle-Free Fluid Transfer System, CODAN Spikes / MicroSpikes
Cannulae
The CODAN Spikes are a compact and safe needle-free system for the manual preparation, withdrawal or transfer of cytostatics and pharmaceuticals as well as their solvent solutions. In order to protect users from aerosols and the drug from micro bacterial contamination, all CODAN Spikes are equipped with a hydrophobic 0,2 µm air filter.
  • Gamma sterilised
  • Cap protects against damage and contamination
  • Integrated, hydrophobic 0,2 µm air filter guarantees sterile aeration and venting and reliably retains aerosols during preparation
  • Minimise the risk of injury
More Product Information