SpectroMembrane® Thin-Film Sample Support Window Frames
Supplier: CHEMPLEX INDUSTRIES
3020 3012 3022-5 3021-5 3024 3090
CHPX3020EA 129 GBP
CHPX3020 CHPX3012 CHPX3022-5 CHPX3021-5 CHPX3024 CHPX3090
SpectroMembrane® Thin-Film Sample Support Window Frames
Sample Containers XRF Sample Cups
SpectroMembrane® Thin-film sample support carrier frames consist of a thin-film sample support substance attached to a frame that serves as a carrier.
  • No waste
  • No wrinkles
  • No static cling
  • No contamination
In affixing a thin-film substance to an XRF Sample Cup, the thin-film material is not directly handled and a contamination possibility is completely eliminated. Nearing or at the completion of attachment, the thin-film automatically detaches from the carrier frame leaving a taut wrinkle-free sample support window.
Etnom® was the first thin-filmMore Product Information