S-Ethylisothiouronium bromide

Supplier: Cayman Chemical

Synonyms: Ethyl carbamimidothioate hydrobromide, S-Ethylisothiourea Hydrobromide, S-EIT hydrobromide, 2-Ethyl-2-thiopseudourea hydrobromide

81275-100 81275-50 81275-500 81275-1
CAYM81275-100EA 50.5 GBP
CAYM81275-100 CAYM81275-50 CAYM81275-500 CAYM81275-1
S-Ethylisothiouronium bromide
S-Ethylisothiouronium bromide
Formula: C₃H₉BrN₂S
MW: 185.1 g/mol
MDL Number: MFCD00012585
CAS Number: 1071-37-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR