Glutathione Peroxidase Assay Kit
Supplier: Cayman Chemical
703102-480
CAYM703102-480EA 951 GBP
CAYM703102-480
Glutathione Peroxidase Assay Kit
Assays Cellular Assays
For the measurement of glutathione-dependent peroxidases in plasma, erythrocyte lysates, tissue homogenates, and cell lysates.
  • Measure GPx in plasma, erythrocyte lysate, tissue homogenates, and cell lysates
  • Assay 45 samples in duplicate
  • Measure GPx activity down to 50 U/ml
  • Plate-based colorimetric measurement (340 nm)
Glutathione peroxidase (GPx) catalyzes the reduction of hydroperoxides, including hydrogen peroxides, by reduced glutathione and functions to protect the cell from oxidative damage. With the exception of phospholipid-hydroperoxide GPx, a monomer, all of the GPx enzymes are tetramers of four identical subunits. Each subunit contains a selenocysteine in the active site which participates directly in the two-electron reduction of the peroxide substrate. The enzyme uses glutathione as the ultimate electron donor to regenerate the reduced form of the selenocysteine. The Cayman Chemical Glutathione Peroxidase Assay Kit measures GPx activity indirectly by a coupled reaction with glutathione reductase (GR). Oxidized glutathione (GSSG), produced upon reduction of an organic hydroperoxide by GPx, is recycled to its reduced state by GR and NADPH. The oxidation of NADPH to NADP+ is accompanied by a decrease in absorbance at 340 nm. The rate of decrease in the A340 is directly proportional to the GPx activity in the sample. The Cayman GPx Assay Kit can be used to measure all of the glutathione-dependent peroxidases in plasma, erythrocyte lysates, tissue homogenates, and cell lysates.