Echinocandin B
Supplier: Cayman Chemical
25520-5 25520-1
CAYM25520-5EA 2060 GBP
CAYM25520-5 CAYM25520-1
Echinocandin B
Proteins and Peptides
Echinocandin B inhibits 1,3-β glucan synthase; inhibits incorporation of radiolabeled glucose into alkali-insoluble glucan by S. cerevisiae spheroplasts; inhibits growth of C. albicans in vitro (MIC = 3.9 µg/ml after 2 days)
  • An echinocandin antifungal
  • Stability: ≥2 years