Tobramycin

Supplier: Cayman Chemical
Danger

Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol

14596-50 14596-100 14596-500
CAYM14596-50EA 54.3 GBP
CAYM14596-50 CAYM14596-100 CAYM14596-500
Tobramycin
Tobramycin
Formula: C₁₈H₃₇N₅O₉
MW: 467.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00077885
CAS Number: 32986-56-4
EINECS: 251-322-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR