Anti-AR Mouse Monoclonal Antibody [clone: 156C135.2]

Supplier: Cayman Chemical
13492-1
CAYM13492-1EA 781 GBP
CAYM13492-1
Anti-AR Mouse Monoclonal Antibody [clone: 156C135.2]
Antibodies
Type: Primary
Antigen: AR
Clonality: Monoclonal
Clone: 156C135.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR