6-Aminonicotinamide

Supplier: Cayman Chemical

Synonyms: 6-Aminopyridine-3-carboxamide

10009315-100 10009315-250 10009315-500 10009315-1
CAYM10009315-100EA 25.3 GBP
CAYM10009315-100 CAYM10009315-250 CAYM10009315-500 CAYM10009315-1
6-Aminonicotinamide
6-Aminonicotinamide
Formula: C₆H₇N₃O
MW: 137.14 g/mol
MDL Number: MFCD00006327
CAS Number: 329-89-5
EINECS: 206-349-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR