Tip cleaning sponge, Hexacon
Supplier: CAULFIELD INDUSTRIAL
SP8141C
CAULSP8141CEA 81.3 GBP
CAULSP8141C
Tip cleaning sponge, Hexacon
Wipes Sponges
This cellulose sponge with one hole provides an effective method to clean soldering residues from a hot tip. With regular use additional abrasive cleaning can become unnecessary.
  • Purpose-made for tip cleaning
  • Large continuous surface area
  • Very low sulphur and acid content prevents tip contamination
  • Fine pore texture suits all tip sizes