Anti-IgG Antibody (HRP (Horseradish Peroxidase))

Supplier: Boster Bio
BA1060-1
BSBTBA1060-1EA 0 GBP
BSBTBA1060-1
Anti-IgG Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Peroxidase
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR