Burettes, lateral stopcock, Schellbach, with USP individual certificate or DAkkS calibration certificate, BLAUBRAND®
Supplier: Brand
USP13844 USP13847 USP13848 DAKKS13848 DAKKS13847 DAKKS13844
BRNDUSP13844EA 173 GBP
BRNDUSP13844 BRNDUSP13847 BRNDUSP13848 BRNDDAKKS13848 BRNDDAKKS13847 BRNDDAKKS13844
Burettes, lateral stopcock, Schellbach, with USP individual certificate or DAkkS calibration certificate, BLAUBRAND®
Burettes
Borosilicate glass 3,3, PTFE spindle, class AS.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Marks and inscriptions in high contrast blue enamel represents the optimum combination of resistance and readability
Length: 800 mm
Length: 800 mm
Certifications: DIN EN ISO 385
Order Now
SPECIFICATIONS
Environmentally Preferable