Counting chambers for CSF, BLAUBRAND® Nageotte
Supplier: Brand
721305
BRND721305EA 156 GBP
BRND721305
Counting chambers for CSF, BLAUBRAND® Nageotte
Slides Counting Chambers
This counting chamber is used for counting cells in the cerebro-spinal fluid (CSF) as well as for counting nematodes.
The square area of 100 mm² is subdivided into 40 rectangles with an area of 2,5 mm² each.