Label Printer, UK with Brady Workstation Suite, M510
Supplier: Brady
322727
BRDY322727EA 489 GBP
BRDY322727
Label Printer, UK with Brady Workstation Suite, M510
Printers Label Printers
The perfect companion for your project workspace, the M510 delivers high-volume, high-quality labels day in and day out. Whether you're in a lab, field or facility, this printer works as hard as you do.
  • Create large 38,10 mm labels with edge-to-edge printing and a wide range of text sizes
  • Print up to 38,10 mm per second to get work done faster
  • Use the M510 for more applications with 250+ hassle-free drop-in cartridges and 26+ high-performance materials
  • Enter data with more accuracy as you work between an intuitive QWERTY keypad, a separate numeric keypad
More Product Information