Fire Retardant Oval Tags for M710 and BMP71
Supplier: Brady
174538
BRDY174538EA 111.98 GBP
BRDY174538
Fire Retardant Oval Tags for M710 and BMP71
Labels
Rigid polyethylene tags (B-145) are printable, non-adhesive, tear-resistant, flame retardent tags used for a wide array of identification applications.
  • Constructed from a 9,5 mil, cross laminated polyethylene film and a thermal transfer printable topcoat
  • Suitable for thermal transfer printing, then folded over and held together in a two-ply format with a pressure sensitive adhesive
  • Operating temperature of −40 to +100 °C (−40 to +212 °F)
  • Matte finish
Applications include asset tracking, component and equipment labelling, data and telecommunicationsMore Product Information